Learn More
R&D Systems™ Recombinant Viral B8R Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand:  R&D Systems™ 8225-BR-025/CF
 View more versions of this product
Product Code. 18757377
  • £275.00 / 25µg
Explore more special offers
Description
Description
The Recombinant Viral B8R Protein is derived from CHO. The Recombinant Viral B8R Protein has been validated for the following applications: Bioactivity.
Order Info
The product is shipped at ambient temperature, upon receipt, store immediately
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS
Chinese Hamster Ovary cell line,CHO-derived viral B8R protein Lys18-Ser272,with a C-terminal 6-His tag
Unconjugated
25 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.