missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Viral B8R Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
£275.00
Specifications
Quantity | 25 μg |
---|---|
Source | Chinese Hamster Ovary cell line,CHO-derived viral B8R protein Lys18-Ser272,with a C-terminal 6-His tag |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Conjugate | Unconjugated |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18757377
|
R&D Systems™
8225-BR-025/CF |
Lyophilized from a 0.2μm filtered solution in PBS |
£275.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18709187
|
R&D Systems™
8225-BR-025 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein |
£275.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Viral B8R Protein is derived from CHO. The Recombinant Viral B8R Protein has been validated for the following applications: Bioactivity.Order Info
The product is shipped at ambient temperature, upon receipt, store immediately
Specifications
25 μg | |
Chinese Hamster Ovary cell line,CHO-derived viral B8R protein Lys18-Ser272,with a C-terminal 6-His tag | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title