Learn More
Abeba™ Reflexor™ 31092 Shoes
ESD dissipative shoe includes PU-sole with high slip resistance.
Brand: Abeba™ 31092/40
Additional Details : Weight : 0.55000kg
Description
- Smooth leather
- Black
- Slip resistance PU outsole sole
- Integrated Reflexor™ massage footbed
- ESD dissipative according to DIN EN 61340
- Lining microfiber, respiration absorbing
- Hand-sewn seam
Specifications
EN 61340 | |
Microfiber | |
40 | |
Polyurethane |
Black | |
Leather | |
Slip-resistant | |
31092 |