Learn More
Abeba™ Reflexor™ 36903 Shoes
ESD dissipative shoe includes PU-sole with high slip resistance.
Brand:  Abeba™ 36903/39
102.15 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Additional Details : Weight : 0.85000kg
 View more versions of this product
Product Code. 15600808
  • £107.53 / 1 pair
Estimated Shipment: 15-08-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, E SRB
  • Smooth leather white
  • Slip resistance PU outsole sole
  • 2 replaceable Reflexor™ insoles (article no. 3565/ 3567)
  • ESD dissipative according to DIN EN 61340
  • Heel strap foldable, adjustable
  • Lining microfiber, respiration absorbing
  • Instep area with elastic band
  • Buckle fastening
  • Hand-sewn seam
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E SRB
Microfiber
39
Polyurethane
White
Leather
Slip-resistant
36903