missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Refrigerant Pack Phase Change Gel
Keep sensitive materials at the right temperature. Returns to gel state when thawed.
Brand: Cole-Parmer™ 06347-76
This item is not returnable.
View return policy
Description
- Phase change formula lasts 15% longer than standard gel packs
- Four formulations from 32 to -10°F (0 to -23°C)
- Nontoxic and reusable
Specifications
19.7 cm | |
7.75 in. | |
1.18 in. | |
48 oz. | |
1419.5 mL | |
-1°C (30°F) |
10.5 in. | |
26.7 cm | |
3 cm | |
1360 g | |
Phase Change Cold Pack, Soft, 30F, 48 oz | |
26.7 x 19.7 x 3 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction