missing translation for 'onlineSavingsMsg'
Learn More
Reinecke Salt, ACS Reagent, Thermo Scientific Chemicals Product Code.: 10790292

Reinecke Salt, ACS Reagent, Thermo Scientific Chemicals

Product Code. 10790292
25 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10790292

Brand: Thermo Scientific Acros 423330250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13573-16-5
Molecular Formula C4H10CrN7S4·H2O
Molecular Weight (g/mol) 354.46
MDL Number MFCD00150925
Synonym Ammonium reineckate, Ammonium tetrathiocyanatodiamminechromate

Specifications

Melting Point 270.0°C
Color Red
Physical Form Crystalline Powder
Quantity 25 g
Linear Formula NH4(Cr(NH3)2(SCN)4)·H2O
Merck Index 15, 8248
Solubility Information Solubility in water: soluble in hot water. Other solubilities: insoluble in deluted hydrochloric acid,soluble in ethanol
Formula Weight 354.46
Grade ACS Reagent
Infrared Spectrum Authentic
Packaging Glass bottle
Sensitivity Passes Test
Chemical Name or Material Reinecke salt
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466445 Certificate TypeCertificate of Analysis Date30/09/2024 Catalog Number423330250,423335000,423331000
Lot NumberA0448132 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number423330250,423335000,423331000
Lot NumberA0454979 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number423330250,423335000,423331000
Lot NumberA0401924 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number423330250,423335000,423331000
Lot NumberA0397913 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number423330250,423335000,423331000
compliance-icons
Product Identifier
  • Reinecke salt
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Reinecke Salt, ACS Reagent, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.