Learn More
Memmert™ Reinforced Stainless Steel Grid
Additional reinforced stainless steel grid for reinforced chamber option
Brand:  Memmert™ IPP750ecoB32190
 View more versions of this product
Product Code. 17120255
  • £478.43 / Each
Estimated Shipment: 23-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Consider max. loading of chamber; electropolished, max. loading 60 kg
Specifications
Specifications
IPP750eco