Learn More
Memmert™ Reinforced Stainless Steel Grid
Additional reinforced stainless steel grid for reinforced chamber option
£241.80 - £561.04
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
17120255
|
Memmert™
IPP750ecoB32190 |
IPP750eco |
£478.43
Each |
Estimated Shipment: 23-12-2024
Log in to see stock. |
|||||
17130105
|
Memmert™
IPP110ecoE29767 |
IPP110eco |
£241.80
Each |
Estimated Shipment: 23-12-2024
Log in to see stock. |
|||||
17139784
|
Memmert™
HPP260ecoE29766 |
HPP260eco |
£326.99
Each |
Estimated Shipment: 23-12-2024
Log in to see stock. |
|||||
17160225
|
Memmert™
IPP410ecoE29766 |
IPP410eco |
£326.99
Each |
Estimated Shipment: 23-12-2024
Log in to see stock. |
|||||
17169854
|
Memmert™
HPP750ecoB32190 |
HPP750eco |
£478.43
Each |
Estimated Shipment: 23-12-2024
Log in to see stock. |
|||||
17170185
|
Memmert™
IPP260ecoE29766 |
IPP260eco |
£326.99
Each |
Estimated Shipment: 23-12-2024
Log in to see stock. |
|||||
17180045
|
Memmert™
IPP1060ecoB32550 |
IPP1060eco |
£561.04
Each |
Estimated Shipment: 23-12-2024
Log in to see stock. |
|||||
17189824
|
Memmert™
HPP410ecoE29766 |
HPP410eco |
£326.99
Each |
Estimated Shipment: 23-12-2024
Log in to see stock. |
|||||
17199624
|
Memmert™
HPP1060ecoB32550 |
HPP1060eco |
£561.04
Each |
Estimated Shipment: 23-12-2024
Log in to see stock. |
|||||
17199684
|
Memmert™
HPP110ecoE29767 |
HPP110eco |
£241.80
Each |
Estimated Shipment: 23-12-2024
Log in to see stock. |
|||||
Description
- Consider max. loading of chamber; electropolished, max. loading 60 kg