Learn More
Memmert™ Reinforced Stainless Steel Grid
Additional reinforced stainless steel grid for reinforced chamber option
Brand:  Memmert™ HPP260ecoE29766
 View more versions of this product
Product Code. 17139784
  • £364.00 / Each
Estimated Shipment: 28-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Consider max. loading of chamber; electropolished, max. loading 60 kg
Specifications
Specifications
HPP260eco