Learn More
Memmert™ Reinforced Stainless Steel Grid
Additional reinforced stainless steel grid for reinforced chamber option
Brand: Memmert™ HPP410ecoE29766
This item is not returnable.
View return policy
Description
- Consider max. loading of chamber; electropolished, max. loading 60 kg
Specifications
HPP410eco |