Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella H
Used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
Brand:  Thermo Scientific™ R30160201
 View more versions of this product
Product Code. 11983332
  • £122.00 / 2mL
Explore more special offers
Description
Description
Remel Agglutinating Sera, Salmonella H is used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
  • Select sera also for use as positive controls for stained Salmonella suspensions
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Positive
Yes
Agglutinating Sera
2 mL
Salmonella H (a)
Salmonella H Antigen
2 mL/Vial
Salmonella