missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella H
Used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
£105.00 - £164.00
Specifications
Control Sets | Positive |
---|---|
Detectable Analytes | Salmonella H Antigen |
Type | Agglutinating Sera |
Packaging Type | 2 mL/Vial |
Quantity | 2 mL |
Product Code | Brand | Description | Detectable Analytes | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Detectable Analytes | Quantity | Price | Quantity & Availability | |||
13245579
|
Thermo Scientific™
R30161601 |
Salmonella H (i) | Salmonella H Antigen | 2 mL |
£118.00
2mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11933342
|
Thermo Scientific™
R30160401 |
Salmonella H © | Salmonella H Antigen | 2 mL |
£116.00
2mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11983332
|
Thermo Scientific™
R30160201 |
Salmonella H (a) | Salmonella H Antigen | 2 mL |
£117.00
2mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11923332
|
Thermo Scientific™
R30161001 |
Salmonella H (G) Polyvalent, for gm, gp, fg, gq, fgt, gms, gmt, gt, gmq, gpu | Salmonella H Antigen | 2 mL |
£164.00
2mL |
Estimated Shipment: 23-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12719290
|
Thermo Scientific™
R30160801 |
Salmonella H (enx) | Salmonella H Antigen | 2 mL |
£116.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11923342
|
Thermo Scientific™
R30160301 |
Salmonella H (b) | Salmonella H Antigen | 2 mL |
£116.00
2mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12749290
|
Thermo Scientific™
R30161101 |
Salmonella H (f, g) | Salmonella H Antigen | 2 mL |
£141.00
Each |
Estimated Shipment: 23-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12789280
|
Thermo Scientific™
R30160501 |
Salmonella H (d) | Salmonella H Antigen | 2 mL |
£116.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12667429
|
Thermo Scientific™
R30163301 |
Salmonella H (1, 2) | Salmonella H Antigen | 2 mL |
£116.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12687429
|
Thermo Scientific™
R30163501 |
Salmonella H (1, 6) | Salmonella H Antigen | 2 mL |
£115.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12759290
|
Thermo Scientific™
R30161201 |
Salmonella H (gm) | Salmonella H Antigen | 2 mL |
£116.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12677499
|
Thermo Scientific™
R30858501 |
Salmonella H Polyvalent, Phases 1 and 2 | Salmonella H Antigen | 2 mL |
£117.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12759300
|
Thermo Scientific™
R30162201 |
Salmonella H™ | Salmonella H Antigen | 2 mL |
£116.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12709290
|
Thermo Scientific™
R30160701 |
Salmonella H (eh) | Salmonella H Antigen | 2 mL |
£115.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12749280
|
Thermo Scientific™
R30160101 |
Salmonella H Rapid Diagnostic 3 (d, E, G, k) | Salmonella H Antigen | 2 mL |
£117.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12719300
|
Thermo Scientific™
R30161801 |
Salmonella H (L) Polyvalent, for lv, lw, lz13, lz28, lz38, lz40 | Salmonella H Antigen | 2 mL |
£115.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12660008
|
Thermo Scientific™
R30162301 |
Salmonella H (y) | Salmonella H Antigen | 2 mL |
£116.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
13265579
|
Thermo Scientific™
R30163201 |
Salmonella H Polyvalent, Phase 2 (1, 2, 5, 6, 7) | Salmonella H Antigen | 2 mL |
£118.00
2mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12697579
|
Thermo Scientific™
R30959801 |
Salmonella H Rapid Diagnostic 1 (b, d, E, r) | Salmonella H Antigen | 2 mL |
£117.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12623658
|
Thermo Scientific™
R30959901 |
Salmonella H Rapid Diagnostic 2 (b, E, k, L) | Salmonella H Antigen | 2 mL |
£117.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12617429
|
Thermo Scientific™
R30162801 |
Salmonella H (z27) | Salmonella H Antigen | 2 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12677429
|
Thermo Scientific™
R30163401 |
Salmonella H (1, 5) | Salmonella H Antigen | 2 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12697429
|
Thermo Scientific™
R30163601 |
Salmonella H (1, 7) | Salmonella H Antigen | 2 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
13235579
|
Thermo Scientific™
R30160601 |
Salmonella H (E) Polyvalent, for eh, enx, enz15 | Salmonella H Antigen | 2 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Beschreibung
Remel Agglutinating Sera, Salmonella H is used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
- Select sera also for use as positive controls for stained Salmonella suspensions
Spezifikation
Positive | |
Agglutinating Sera | |
2 mL |
Salmonella H Antigen | |
2 mL/Vial |