Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella H
Used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
Brand:  Thermo Scientific™ R30160101
 View more versions of this product
Product Code. 12749280
  • £122.00 / Each
Explore more special offers
Description
Description
Remel Agglutinating Sera, Salmonella H is used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
  • Select sera also for use as positive controls for stained Salmonella suspensions
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Positive
Salmonella H Antigen
2 mL/Vial
Salmonella H
Salmonella H Rapid Diagnostic 3 (d, E, G, k)
Agglutinating Sera
2 mL