Learn More
Thermo Scientific™ Agglutinating Sera, Salmonella O/Vi
Serologically identify somatic O/Vi antigens of Salmonella strains by slide and/or tube agglutination test using Agglutinating Sera, Salmonella O/Vi.
Brand: Thermo Scientific™ R30957401
Code : 50
Additional Details : Weight : 0.30000kg
Description
Serologically identify somatic O/Vi antigens of Salmonella strains by slide and/or tube agglutination test for epidemiological and diagnostic purposes using Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella O/Vi. The sera are absorbed to remove agglutinins for other Salmonella antigens and the ‘paracolon’ α agglutinin. Salmonella Somatic Agglutinating Sera (ZC series) are intended for the identification of O/Vi antigens.
Serological tests are based on the fact that antibodies in serum, produced in response to exposure to bacterial antigens, will agglutinate with bacteria carrying homologous antigens.
The genus Salmonella is classified in the Kauffmann-White Scheme into serotypes according to combinations of somatic (O) and flagellar (H) antigens, which are identified by agglutination tests1. Agglutinating sera should be used in confirmatory tests but may also be used with appropriate caution in screening tests2. These simple tests rapidly detect specific bacterial antigens for the identification, grouping and/or typing of Salmonella and other food-borne microorganisms. This incorporates agglutinating sera for the identification somatic O/Vi antigens of Salmonella (groups A-S), allowing the fast, accurate and reliable identification of Salmonella strains.
- Ready-to-use: Liquid reagents in 2mL vials
- Rapid: Results within 1 minute.
- Easy-to-interpret: Visible agglutination indicates a positive result
- Less sample required
Each kit contains one bottle of antisera (2mL) preserved either with 0.5% phenol or 0.1% sodium azide. The sera are produced in rabbits.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Positive | |
Salmonella O Antigen | |
Polyvalent | |
2 mL |
Salmonella O/Vi | |
Agglutinating Sera | |
2 mL/Vial | |
Salmonella O/Vi |