Learn More
Thermo Scientific™ Remel™ Contact Plate Sterile Sabouraud Dextrose Agar w/Lecithin, Polysorbate 80 (pH 5.6) (Irradiated)
Thermo Scientific™ Remel Contact Plate Sterile Sabouraud Dextrose Agar with Lecithin, Polysorbate 80 (pH 5.6, Irradiated) is used for cultivation of yeast and fungi.
Brand:  Thermo Scientific™ R111805
118.70 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 17702648
  • £124.00 / Pack of 10
Estimated Shipment: 24-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Thermo Scientific™ Remel Contact Plate Sterile Sabouraud Dextrose Agar with Lecithin, Polysorbate 80 (pH 5.6) (Irradiated) is used for the cultivation of yeast and fungi.
  • Contact Plates Sterile are double bagged plus a 3rd bag is enclosed for customer use (Double + Bag)
  • These products are manufactured in a clean room and validated sterile by gamma radiation
  • The Contact Plates Sterile listed are made using GripFit™ technology, which ensures the lid remains in place during handling and yet can be easily removed during environmental monitoring
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Contact Plates; Sabouraud Dextrose Agar w/Lecithin; Gamma irradiated
Contact - Double Bag
Agar
10/Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Remel™ Contact Plate Sterile Sabouraud Dextrose Agar w/Lecithin, Polysorbate 80 (pH 5.6) (Irradiated) > Contact Plates; Sabouraud Dextrose Agar w/Lecithin; Gamma irradiated;, 10/pk
Not intended for IVD use.