Learn More
Thermo Scientific™ Remel™ Contact Plate Sterile Sabouraud Dextrose Agar w/Lecithin, Polysorbate 80 (pH 5.6) (Irradiated)
Thermo Scientific™ Remel Contact Plate Sterile Sabouraud Dextrose Agar with Lecithin, Polysorbate 80 (pH 5.6, Irradiated) is used for cultivation of yeast and fungi.
£124.00
Specifications
Description Contact Plates; Sabouraud Dextrose Agar w/Lecithin; Gamma irradiated
Format Contact - Double Bag
Product Type Agar
View More Specs
Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17702648
View Documents
Thermo Scientific™
R111805
10/Pk.
£124.00
Pack of 10
Estimated Shipment: 24-04-2025
to see stock.
Add to basket
Description
Description
Thermo Scientific™ Remel Contact Plate Sterile Sabouraud Dextrose Agar with Lecithin, Polysorbate 80 (pH 5.6) (Irradiated) is used for the cultivation of yeast and fungi.
  • Contact Plates Sterile are double bagged plus a 3rd bag is enclosed for customer use (Double + Bag)
  • These products are manufactured in a clean room and validated sterile by gamma radiation
  • The Contact Plates Sterile listed are made using GripFit™ technology, which ensures the lid remains in place during handling and yet can be easily removed during environmental monitoring
Specifications
Specifications
Contact Plates; Sabouraud Dextrose Agar w/Lecithin; Gamma irradiated
Contact - Double Bag
Agar
Videos
SDS
Documents
Documents
Product Certifications
Not intended for IVD use.
Product Content Correction
Product Title
Thermo Scientific™ Remel™ Contact Plate Sterile Sabouraud Dextrose Agar w/Lecithin, Polysorbate 80 (pH 5.6) (Irradiated)