Learn More
Thermo Scientific™ Remel™ MacConkey Agar
The Thermo Scientific™ Remel MacConkey Agar is used for selective and differential isolation of gram-negative bacilli on the basis of lactose fermentation.
£161.00
Specifications
Description MacConkey Agar
Form Powder
Product Type Agar
View More Specs
Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
12647619
View Documents
Thermo Scientific™
R453802
500 g
£161.00
500g
Description
Description
Thermo Scientific™ Remel MacConkey Agar is used for selective and differential isolation of gram-negative bacilli on the basis of lactose fermentation.
Specifications
Specifications
MacConkey Agar
Powder
Agar
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Remel™ MacConkey Agar