Learn More
Thermo Scientific™ Remel™ MacConkey Agar
The Thermo Scientific™ Remel MacConkey Agar is used for selective and differential isolation of gram-negative bacilli on the basis of lactose fermentation.
Brand: Thermo Scientific™ R453802
154.07 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Thermo Scientific™ Remel MacConkey Agar is used for selective and differential isolation of gram-negative bacilli on the basis of lactose fermentation.

Specifications
Industrial Reference: BAM, COMPF, SMWW, USP. | |
Powder | |
500 g |
MacConkey Agar | |
Agar |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.