Learn More
Thermo Scientific™ Remel™ PathoDX™ RSV Control Slide
Remel PathoDx and RSV Control Slide is used with RSV immunofluorescent assay.
Brand:  Thermo Scientific™ R62375
Product Code. 12761780
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Remel PathoDx and RSV Control Slide is used with RSV immunofluorescent assay.
Ordering Alerts:
Each slide has 1 positive and 1 negative well.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PathoDx™ RSV Control Slides
Control
5 Slides/Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3462187 Certificate TypeCertificate of Analysis Date27/05/2022 Catalog NumberR62375
Lot Number3343065 Certificate TypeCertificate of Analysis Date03/03/2022 Catalog NumberR62375
Lot Number3210263 Certificate TypeCertificate of Analysis Date12/02/2021 Catalog NumberR62375
Lot Number3139697 Certificate TypeCertificate of Analysis Date28/01/2021 Catalog NumberR62375
Lot Number2935017 Certificate TypeCertificate of Analysis Date07/12/2020 Catalog NumberR62375
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Remel™ PathoDX™ RSV Control Slide > PathoDx™ RSV Control Slides