Learn More
Thermo Scientific™ Remel™ Rappaport-Vassiliadis Salmonella Enrichment Broth
Remel Rappaport-Vassiliadis Salmonella Enrichment Broth is used for the selective enrichment and isolation of Salmonella species.
Brand: Thermo Scientific™ R455432
205.22 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Remel Rappaport-Vassiliadis Salmonella Enrichment Broth is used for the selective enrichment and isolation of Salmonella species.
Ordering Alerts:
Industrial Reference: USP.

Specifications
Conforms to USP standards | |
Powder | |
500 g |
Rappaport-Vassiliadis Salmonella Enrichment Broth | |
Dehydrated Culture Media |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Laboratory Use Only