Learn More
Corning™ Step-R™ Repeating Pipettor Syringe Tips, Sterile
Corning™ Step-R™ Repeating Pipettor Syringe Tips, Sterile provide precise, positive-displacement dispensing of viscous or volatile liquids.
Brand:  Corning™ 6634
Additional Details : Weight : 5.00000kg
 View more versions of this product
Product Code. 15883905
  • £31.66 / Pack of 25
Estimated Shipment: 11-12-2024
to see stock.
Add to basket
Description
Description
  • Ideal for viscous, dense and volatile liquides
  • Minimum increments printed on the syringe itself
  • Accurately deliver selected dose
  • Made from high quality polypropylene and polyethylene
  • Pyrogen-, ATP-, RNase-, and DNase-free
Specifications
Specifications
Autoclavable
Polypropylene
Dispensing of viscous and volatile liquids
25 mL
ISO8655 ISO11137
Sterile
25 mL