Learn More
Grant Instruments™ Replacement ABS Lid for Grant Ultrasonic Baths
Reduce operating noise and potential escape of aerosols. Grant Instruments™ Replacement ABS Lid for Grant Ultrasonic Baths are supplied as standard with the bath.
Brand:  Grant Instruments™ XAL18
Code : 57
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 11441008
  • £61.00 / Each
Estimated Shipment: 22-08-2024
to see stock.
Add to basket
Description
Description
  • ABS construction
  • For use with XUB and XUBA Series Ultrasonic Baths
Specifications
Specifications
XAB18 Basket for Ultrasonic Bath
ABS
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products