Learn More
Grant Instruments™ Replacement ABS Lid for Grant Ultrasonic Baths
Reduce operating noise and potential escape of aerosols. Grant Instruments™ Replacement ABS Lid for Grant Ultrasonic Baths are supplied as standard with the bath.
£39.00 - £63.00
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
11431008
|
Grant Instruments™
XAL12 |
XAB12 Basket for Ultrasonic Bath |
£53.00
Each |
Estimated Shipment: 22-08-2024
Log in to see stock. |
|||||
11421008
|
Grant Instruments™
XAL10 |
XAB10 Basket for Ultrasonic Bath |
£57.00
Each |
Estimated Shipment: 22-08-2024
Log in to see stock. |
|||||
11441008
|
Grant Instruments™
XAL18 |
XAB18 Basket for Ultrasonic Bath |
£61.00
Each |
Estimated Shipment: 22-08-2024
Log in to see stock. |
|||||
11451008
|
Grant Instruments™
XAL25 |
XAB25 Basket for Ultrasonic Bath |
£63.00
Each |
Estimated Shipment: 22-08-2024
Log in to see stock. |
|||||
11411008
|
Grant Instruments™
XAL5 |
XAB5 Basket for Ultrasonic Bath |
£46.00
Each |
Estimated Shipment: 22-08-2024
Log in to see stock. |
|||||
11490998
|
Grant Instruments™
XAL1 |
XABA1 Basket for Ultrasonic Bath |
£39.00
Each |
Estimated Shipment: 22-08-2024
Log in to see stock. |
|||||
11401008
|
Grant Instruments™
XAL3 |
XABA3 Basket for Ultrasonic Bath |
£41.00
Each |
Estimated Shipment: 22-08-2024
Log in to see stock. |
|||||
Description
- ABS construction
- For use with XUB and XUBA Series Ultrasonic Baths
Specifications
ABS |