Learn More
Grant Instruments™ Replacement Base Tray for Unstirred Water Baths
Required if flat-bottomed flasks are to be placed directly on the base of the bath. Grant Instruments Replacement Base Tray for Unstirred Water Baths promote thermal convection in the bath.
Brand:  Grant Instruments™ AQBT26
 View more versions of this product
Product Code. 11446417
  • £73.10 / Each
Estimated Shipment: 30-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with Sub Aqua Pro Series Unstirred Water Bath
  • Choose a rack that fits 2, 5, 12, or 26L tank size
  • Racks are supplied as standard with purchase of a Sub Aqua Pro Unstirred water Bath
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sub Aqua Pro 18/26L
Base Tray
Polycarbonate
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Grant Instruments™ Replacement Base Tray for Unstirred Water Baths > For Use With: SAP18/26