Learn More
Grant Instruments™ Replacement Basket for Grant Ultrasonic Baths
Designed specifically to generate maximum ultrasonic activity. Grant Instruments™ Replacement Basket for Grant Ultrasonic Baths prevent operators coming into contact with chemical solutions.
Brand:  Grant Instruments™ XAB12
 View more versions of this product
Product Code. 11401018
  • £179.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Stainless steel construction
  • Works in conjunction with lid to create a drip collection unit
  • Prevents user from coming into contact with chemical solutions
  • Prevents items from resting on the inside of the tank wall
  • For use with Grant Ultrasonic Baths
Specifications
Specifications
XUB12 Ultrasonic Bath
Stainless Steel
(WxDxH) 263 x 203 x 193 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products