Learn More
Grant Instruments™ Replacement Basket for Grant Ultrasonic Baths
Designed specifically to generate maximum ultrasonic activity. Grant Instruments™ Replacement Basket for Grant Ultrasonic Baths prevent operators coming into contact with chemical solutions.
£49.00 - £165.00
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
11491008
|
Grant Instruments™
XAB10 |
XUB10 Ultrasonic Bath |
£101.00
Each |
Estimated Shipment: 22-08-2024
Log in to see stock. |
|||||
11401018
|
Grant Instruments™
XAB12 |
XUB12 Ultrasonic Bath |
£118.00
Each |
In Stock
Log in to see stock. |
|||||
11411018
|
Grant Instruments™
XAB18 |
XUB18 Ultrasonic Bath |
£150.00
Each |
Estimated Shipment: 24-09-2024
Log in to see stock. |
|||||
11421018
|
Grant Instruments™
XAB25 |
XUB25 Ultrasonic Bath |
£165.00
Each |
Estimated Shipment: 24-09-2024
Log in to see stock. |
|||||
11481008
|
Grant Instruments™
XAB5 |
XUB5 Ultrasonic Bath |
£77.00
Each |
Estimated Shipment: 22-08-2024
Log in to see stock. |
|||||
11461008
|
Grant Instruments™
XAB1 |
XUBA1 Ultrasonic Bath |
£49.00
Each |
Estimated Shipment: 24-09-2024
Log in to see stock. |
|||||
11471008
|
Grant Instruments™
XAB3 |
XUBA3 Ultrasonic Bath |
£62.00
Each |
Estimated Shipment: 24-09-2024
Log in to see stock. |
|||||
Description
- Stainless steel construction
- Works in conjunction with lid to create a drip collection unit
- Prevents user from coming into contact with chemical solutions
- Prevents items from resting on the inside of the tank wall
- For use with Grant Ultrasonic Baths
Specifications
Stainless Steel |