Learn More
Electrothermal™ Replacement Digi-Mantle Heater Cartridge
Enables a round bottom flask to be stirred and heated up to an element temperature of 450°C. Electrothermal™ Replacement Digi-Mantle Heater Cartridge uses precise electronic control to provide accurate e of temperature and stirring measures.
Brand:  Electrothermal™ 36204-01
 View more versions of this product
Product Code. 11757873
  • £825.00 / Each
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with the OMCA Digital Controller.
  • Stir and heat round bottom flasks up to an element temperature of 450°C
  • Uses precise electronic control to give an accurate measure of temperature and stirring.
  • Modular design allows heating cartridges of different sizes to be interchanged using the same OMCA digital controller as a base
  • Cost-effective and versatile
Specifications
Specifications
250 mL
OMCA Digi-Mantle
Replacement Heating Cartridge