Learn More
Thermo Scientific™ Replacement Needles for GC Syringes
Thermo Scientific™ Replacement Needles for Syringes are available for syringes with removable needles.
Brand: Thermo Scientific™ 36550046
79.09 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Thermo Scientific™ Replacement Needles for Syringes are available for syringes with removable needles.
Specifications
36550046 | |
Replacement Syringe Needle | |
75 mm | |
Thermo Scientific 36500520 Syringe |
31 G | |
10 μL | |
Cone | |
Removable |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.