missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BSTFA + 1–10% TMCS Silylation Reagent

Ideal for difficult-to-silylate compounds, the Thermo Scientific™ BSTFA + 1–10% TMCS Silylation Reagent is derivatizing compounds.

Brand:  Thermo Scientific™ TS-38840

276.24 GBP valid until 2025-04-30
Use promo code "24199" to get your promotional price.



 View more versions of this product

Product Code. 11577841

  • £305.91 / 10mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Use Thermo Scientific™ BSTFA + 1–10% TMCS Silylation Reagent for analytes that are difficult to silylate. Stronger than BSTFA alone, this catalyzed formulation is ideal for derivatizing compounds such as fatty acid amides and slightly hindered hydroxyls for GC analysis.

  • Excellent for derivatizing fatty acid amides, slightly hindered hydroxyls and other compounds
  • Catalyzed formulation is stronger than BSTFA alone
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BSTFA +10% TMCS; 10 x 1 mL Ampules
2°C to 8°C
10 x 1 mL Ampules
BSTFA +10% TMCS
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

  • Multiple Manufacturers
    Take advantage of our exclusive monthly promotions on high-quality chemicals.
    Promo Code: 24199

    Expires2025-04-30

    Days

    26

    Hours

    22

    Minutes

    56

    Seconds

    9
  • Multiple Manufacturers
    Explore our new promotion and get up to 70% off some of our most popular products.
    Promo Code: 24127

    Expires2025-05-31

    Days

    57

    Hours

    22

    Minutes

    56

    Seconds

    9
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ BSTFA + 1–10% TMCS Silylation Reagent > BSTFA +10% TMCS; 10 x 1mL ampules

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.