missing translation for 'onlineSavingsMsg'
Learn More

Alla France™ Researcher Alcoholmeter

Alla France™ Researcher Alcoholmeter is used to determine the amount of alcohol in a liquid.

Brand:  Alla France™ 0530FC070/20-QP

 View more versions of this product

Product Code. 15974412

  • £26.40 / Each
Estimated Shipment: 10-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Calibration temperature 20°C
  • High precision
  • For use in the distillery and wine industry
  • Includes thermometer
  • With fiducial mark
  • Reading below meniscus
  • With individual number
Specifications

Specifications

Alcohol Hydrometer
0.005
Certificate of Conformity
Glass
±0.5%
320 mm
70 to 100% Vol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers