Learn More
Thermo Scientific™ Reservoir Accessories
Level control and sterile overflow for storage reservoirs are perfect accessories for running reverse osmosis systems.
Brand:  Thermo Scientific™ 06.5560
 View more versions of this product
Product Code. 15514780
  • £1703.66 / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Level control and sterile overflow, Thermo Scientific™ Reservoir Accessories, are perfect for running reverse osmosis systems.
Level Control for Storage Reservoirs
  • Required accessory for running any reverse osmosis system; will be adapted to the specific reservoir during the order procedure
Sterile Overflow for Storage Reservoirs
  • Recommended accessory to prevent organic contamination inside of reverse osmosis reservoirs
Vent filter for Storage Reservoirs
  • Recommended accessory to prevent organic contamination inside of reverse osmosis reservoirs
Vent filter with CO2 absorber for Storage Reservoirs
  • Recommended accessory to prevent organic and CO2 contamination inside of reverse osmosis reservoirs
Ordering Information:
This product is not available in all regions
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Vent filter/CO2 Adsorber and Sterile overflow for tank 1100 - 2000 liters
Reservoir, 1100 to 2000 L
Reservoir Accessory