missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Reservoir Accessories

Level control and sterile overflow for storage reservoirs are perfect accessories for running reverse osmosis systems.

£105.10 - £1703.66


Products 3
Product Code Brand Description For Use With (Equipment) Price Quantity & Availability  
Product Code Brand Description For Use With (Equipment) Price Quantity & Availability  
11347171
View Documents
Thermo Scientific™
22.0091
Sterile Reservoir Vent Filter for Storage tanks, 0.2 μm Smart2Pure Systems, 3 and 6 L/hr.
£105.10
Each
In Stock
to see stock.
Add to basket
 
15172296
View Documents
Thermo Scientific™
50135142
Sterile Reservoir Vent Filter for Storage tank 30 - 100 litres, 0.2 μm LabTower RO
£255.13
Each
In Stock
to see stock.
Add to basket
 
15514780
View Documents
Thermo Scientific™
06.5560
Vent filter/CO2 Adsorber and Sterile overflow for tank 1100 - 2000 liters Reservoir, 1100 to 2000 L
£1703.66
Each
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
Description

Description

Level control and sterile overflow, Thermo Scientific™ Reservoir Accessories, are perfect for running reverse osmosis systems.

Level Control for Storage Reservoirs
  • Required accessory for running any reverse osmosis system; will be adapted to the specific reservoir during the order procedure
Sterile Overflow for Storage Reservoirs
  • Recommended accessory to prevent organic contamination inside of reverse osmosis reservoirs
Vent filter for Storage Reservoirs
  • Recommended accessory to prevent organic contamination inside of reverse osmosis reservoirs
Vent filter with CO2 absorber for Storage Reservoirs
  • Recommended accessory to prevent organic and CO2 contamination inside of reverse osmosis reservoirs
Ordering Information:

This product is not available in all regions
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.