missing translation for 'onlineSavingsMsg'
Learn More
Resorcinol, 98%, Thermo Scientific Chemicals Product Code.: 10149831

Resorcinol, 98%, Thermo Scientific Chemicals

Product Code. 10149831
50 g, Plastic bottle
Click to view available options
Quantity:
50 g
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10149831

Brand: Thermo Scientific Acros 132290500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-46-3
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.11
MDL Number MFCD00002269
InChI Key GHMLBKRAJCXXBS-UHFFFAOYSA-N
Synonym resorcinol, 1,3-benzenediol, resorcin, 1,3-dihydroxybenzene, 3-hydroxyphenol, m-hydroquinone, resorcine, m-dihydroxybenzene, m-hydroxyphenol, m-benzenediolShow MoreShow Less
PubChem CID 5054
ChEBI CHEBI:27810
IUPAC Name benzene-1,3-diol
SMILES C1=CC(=CC(=C1)O)O

Specifications

Melting Point 109.0°C to 111.0°C
Color Colorless to Yellow
Boiling Point 281.0°C
Flash Point 127°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Quantity 50 g
Beilstein 06,II,802
Merck Index 15,8276
Solubility Information Solubility in water: 140 g/100 mL. Other solubilities: freely soluble in ether, glycerol, soluble in benzene and amyl alcohol, slightly soluble in chloroform
Formula Weight 110.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Resorcinol, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464898 Certificate TypeCertificate of Analysis Date02/08/2024 Catalog Number132292500,132290500,132290050,132290010
Lot NumberA0460294 Certificate TypeCertificate of Analysis Date16/02/2024 Catalog Number132292500,132290500,132290050,132290010
Lot NumberA0393534 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number132292500,132290500,132290050,132290010
Lot NumberA0412933 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number132292500,132290500,132290050,132290010
Lot NumberA0415245 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number132292500,132290500,132290050,132290010
compliance-icons compliance-icons
Product Identifier
  • Resorcinol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Resorcinol, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.