missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva RESOURCE™ Q IEX Columns
Use for intermediate lab-scale purification and large-scale polishing of biomolecules Cytiva Life Sciences™ RESOURCE™ Q IEX Columns are strong anion exchanger columns.
£744.64 - £2115.00
Specifications
Kit Contents | Connectors for high- and low-pressure chromatography equipment and instructions |
---|---|
Column Material | PEEK |
Matrix | Polystyrene/divinylbenzene |
For Use With (Application) | RESOURCE Q columns are prepacked with SOURCE 15Q, a strong anion exchanger for high resolution polishing purification of proteins at high flow rates. |
Ligand Type | Quaternary ammonium |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
10479944
|
Cytiva
17-1177-01 |
1 mL |
£744.64
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11564975
|
Cytiva
17-1179-01 |
6 mL |
£2115.00
6mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Rigid, monodispersed, spherical particles with controlled pore-size distribution offer excellent flow characteristics, sample loading of up to 25mg/mL and stability in organic solvents and pH extremes
- Resource columns with 1mL bed volume have separation times of 3 minutes or less at 9.6mL/min. and approximately 20 min. using a peristaltic pump at 1mL/min.
Specifications
Connectors for high- and low-pressure chromatography equipment and instructions | |
Polystyrene/divinylbenzene | |
Quaternary ammonium | |
15 μm |
PEEK | |
RESOURCE Q columns are prepacked with SOURCE 15Q, a strong anion exchanger for high resolution polishing purification of proteins at high flow rates. | |
4°C to 30°C in 20% Ethanol | |
2 to 12 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva RESOURCE™ Q IEX Columns