missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva RESOURCE™ and SOURCE™ 15 and 5RPC columns
Designed for rapid screening experiments, method development and small-scale protein purification using reversed phase chromatography
£702.00 - £926.00
Specifications
Diameter (Metric) | 6.4 mm |
---|---|
Flow Rate | 10 mL/min |
Ligand Type | Matrix surface |
Max. Pressure | 580 psi |
Particle Shape | Monosized |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
10249434
|
Cytiva
17-1181-01 |
1 mL |
£702.00
1mL |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||||
11590804
|
Cytiva
17-1182-01 |
3 mL |
£926.00
3mL |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||||
Description
- Wide pH range (1-12), outstanding selectivity, chemical resistance, high capacity and high resolution at high flow rates
- Excellent scalability from RESOURCE to FineLINE columns
- Use RESOURCE RPC 3 ml column for higher resolution/method development on a 10 cm bed height
Specifications
6.4 mm | |
Matrix surface | |
Monosized | |
1 to 12 | |
No | |
Reversed Phase Column |
10 mL/min | |
580 psi | |
15 μm | |
Polystyrene/Divinyl Benzene | |
PEEK |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- RESOURCE RPC, 1 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.