Learn More
Restek™ 20 mm Aluminum Seals with Septa, Assembled
Preassembled seals save time and increase productivity
Brand:  Restek™ 21761
Code : Z3
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 13129662
  • £114.95 / Pack of 100
Estimated Shipment: 07-10-2024
to see stock.
Add to basket
Description
Description
  • Color: Silver
  • Material: PTFE/Gray Butyl Rubber Septa
Specifications
Specifications
Silver
PTFE, Butyl Rubber
Crimp Cap
Aluminum
100
20 mm