Learn More
Restek™ Accuform Micro-Vials with Screw Threads
Work with Mininert™ sampling valves
Brand: Restek™ 21050
This item is not returnable.
View return policy
Description
- Tapered for high recovery of contents
- With graduated marking spot
- Material: Borosilicate glass
Specifications
Borosilicate Glass | |
Mininert™ sampling valves | |
Tapered | |
12 Pk. | |
Yes |
1 mL | |
Yes | |
Screw Cap (Open-Top) | |
13-425 |