Learn More
Restek™ Glass Vial Inserts
Specially designed for 2 mL, 11 mm crimp top and 2 mL, 9 mm short cap, screw thread vials
£42.35 - £1433.85
Product Code | Brand | Volume (Metric) | Style | For Use With (Equipment) | No. per Pack | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Style | For Use With (Equipment) | No. per Pack | Price | Quantity & Availability | ||
13179672
|
Restek™
21776 |
250 μL | Bottom Spring | Big Mouth Step Design Vials | 100 |
£160.93
Pack of 100 |
Estimated Shipment: 11-09-2024
Log in to see stock. |
||
13189672
|
Restek™
21777 |
250 μL | Bottom Spring | Big Mouth Step Design Vials | 1000 |
£1433.85
Pack of 1000 |
Estimated Shipment: 24-09-2024
Log in to see stock. |
||
13169672
|
Restek™
21775 |
100 μL | Bottom Spring | Standard Mouth Vials | 1000 |
£880.88
Pack of 1000 |
Estimated Shipment: 24-09-2024
Log in to see stock. |
||
13199672
|
Restek™
21779 |
250 μL | Glass Flange (step) | Big Mouth Step Design Vials | 1000 |
£1197.90
Pack of 1000 |
Estimated Shipment: 24-09-2024
Log in to see stock. |
||
13109682
|
Restek™
21780 |
350 μL | Flat Bottom | Big Mouth Step Design Vials | 100 |
£42.35
Pack of 100 |
Estimated Shipment: 16-09-2024
Log in to see stock. |
||
13139682
|
Restek™
21782 |
50 μL | Bottom Spring | Big Mouth Step Design Vials | 1000 |
£876.04
Pack of 1000 |
Estimated Shipment: 24-09-2024
Log in to see stock. |
||
15152875
|
Restek™
24513 |
50 μL | Bottom Spring | Big Mouth Step Design Vials | 100 |
£101.64
Pack of 100 |
Estimated Shipment: 24-09-2024
Log in to see stock. |
||
Description
- Limited-volume inserts also available
- Material: glass
Specifications
Glass |