missing translation for 'onlineSavingsMsg'
Learn More

Restek™ High-capacity and ECD Vent Traps

Includes connecting lines and mounting kit.

Brand:  Restek™ 20698

Code : Z3

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12912609

  • £82.28 / Each
Estimated Shipment: 27-08-2024
to see stock.

Description

Description

  • High-Capacity Split Vent Trap reduces the release of hazardous materials from the capillary split vent into the lab
  • ECD Vent Trap reduces the release of hazardous materials from the ECD vent into the lab
  • Fittings: 1/8 in.
  • Dimensions: 15.2 x 2.5cm (6 x 1 in.)
Specifications

Specifications

0.12 in. Fitting
Connecting lines and mounting kit
6 in.
1 in.
High Capacity Split Vent Trap
1 Each
15.2 cm
2.5 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers