missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Restek™ High-capacity and ECD Vent Traps
Includes connecting lines and mounting kit.
£82.28 - £302.50
Specifications
Connection Type | 0.12 in. Fitting |
---|---|
Includes | Connecting lines and mounting kit |
Length (English) | 6 in. |
Length (Metric) | 15.2 cm |
Diameter (English) Outer | 1 in. |
Product Code | Brand | Type | Packaging Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Packaging Quantity | Price | Quantity & Availability | ||||
12912609
|
Restek™
20698 |
High Capacity Split Vent Trap | 1 Each |
£82.28
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13161412
|
Restek™
22017 |
ECD Vent Trap | 1 Each |
£84.70
Each |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12922609
|
Restek™
20699 |
High Capacity Split Vent Trap | 5/Pk. |
£302.50
Pack of 5 |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- High-Capacity Split Vent Trap reduces the release of hazardous materials from the capillary split vent into the lab
- ECD Vent Trap reduces the release of hazardous materials from the ECD vent into the lab
- Fittings: 1/8 in.
- Dimensions: 15.2 x 2.5cm (6 x 1 in.)
Specifications
0.12 in. Fitting | |
6 in. | |
1 in. |
Connecting lines and mounting kit | |
15.2 cm | |
2.5 cm |