Learn More
Restek™ Replacement Split/Splitless Injection Ports: Replacement Weldments
Replacement weldments for Agilent 5890 and 6890/6850 GCs
Brand:  Restek™ 20266
Encompass_Preferred
 View more versions of this product
Product Code. 13011680
  • £505.78 / Each
Explore more special offers
Description
Description
  • Siltek™ treatment passivates metal surface ensuring an inert pathway for the sample, delivering increased performance
  • High-quality stainless steel injector
  • Meets or exceeds Agilent original equipment performance
Specifications
Specifications
Gas Chromatography
Agilent 5890 GCs
Stainless Steel
Shell Weldment