Learn More
Restek™ Replacement Split/Splitless Injection Ports: Replacement Weldments
Replacement weldments for Agilent 5890 and 6890/6850 GCs
£448.91 - £629.20
Product Code | Brand | For Use With (Equipment) | Material | Product Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Material | Product Type | Price | Quantity & Availability | |||
13001680
|
Restek™
20265 |
Manual Flow or Epc on Agilent 5890 GCs; Manual Flow Only on Agilent 6890/6850 GCs | Stainless Steel | Weldment |
£479.16
Each |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
13011680
|
Restek™
20266 |
Agilent 5890 GCs | Stainless Steel | Shell Weldment |
£505.78
Each |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
13021680
|
Restek™
20267 |
Manual Flow or Epc on Agilent 5890 GCs; Manual Flow Only on Agilent 6890/6850 GCs | Siltek Treated Stainless Steel | Weldment |
£574.75
Each |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
13031680
|
Restek™
20268 |
Agilent 5890 GCs | Siltek Treated Stainless Steel | Shell Weldment |
£578.38
Each |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
13157132
|
Restek™
22673 |
Agilent 6890/6850 GCs | Stainless Steel | Shell Weldmen |
£448.91
Each |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
13137132
|
Restek™
22671 |
Agilent 6890/6850 GCs | Siltek Treated Stainless Steel | Shell Weldment |
£534.82
Each |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
13147132
|
Restek™
22672 |
Agilent 6890/6850 GCs | Siltek Treated Stainless Steel | Weldment with EPC |
£629.20
Each |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
13167132
|
Restek™
22674 |
Agilent 6890/6850 GCs | Stainless Steel | Weldment with EPC |
£592.90
Each |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Siltek™ treatment passivates metal surface ensuring an inert pathway for the sample, delivering increased performance
- High-quality stainless steel injector
- Meets or exceeds Agilent original equipment performance
Specifications
Gas Chromatography |