Learn More
Restek™ Replacement Split/Splitless Injection Ports: Replacement Weldments
Replacement weldments for Agilent 5890 and 6890/6850 GCs
Brand: Restek™ 22672
Code : Z3
Additional Details : Weight : 1.00000kg
Description
- Siltek™ treatment passivates metal surface ensuring an inert pathway for the sample, delivering increased performance
- High-quality stainless steel injector
- Meets or exceeds Agilent original equipment performance
Specifications
Gas Chromatography | |
Weldment with EPC | |
Siltek Treated Stainless Steel | |
Agilent 6890/6850 GCs |