Learn More
Restek™ Rtx™-502.2 Capillary Columns
Application-specific columns with unique selectivity for volatile organics
£739.31 - £1752.08
Product Code | Brand | Length (Metric) | Diameter (Metric) Inner | Film Thickness | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) Inner | Film Thickness | Price | Quantity & Availability | |||
12915539
|
Restek™
10910 |
105 m | 0.53 mm | 3 μm |
£1752.08
Each |
Estimated Shipment: 16-12-2024
Log in to see stock. |
|||
12985529
|
Restek™
10909 |
60 m | 0.53 mm | 3 μm |
£1353.99
Each |
Estimated Shipment: 16-12-2024
Log in to see stock. |
|||
12945539
|
Restek™
10915 |
30 m | 0.25 mm | 1.4 μm |
£739.31
Each |
Estimated Shipment: 16-12-2024
Log in to see stock. |
|||
12995539
|
Restek™
10919 |
30 m | 0.32 mm | 1.8 μm |
£786.50
Each |
Estimated Shipment: 16-12-2024
Log in to see stock. |
|||
10697244
|
Restek™
10916 |
60 m | 0.25 mm | 1.4 μm |
£1223.31
Each |
Estimated Shipment: 16-12-2024
Log in to see stock. |
|||
12965539
|
Restek™
10916 |
60 m | 0.25 mm | 1.4 μm |
£1223.31
Each |
Estimated Shipment: 16-12-2024
Log in to see stock. |
|||
Description
- Proprietary Crossbond™ diphenyl/dimethyl polysiloxane phase
- Selectivity for volatile organic pollutants per US EPA Method 502.2, gasoline range organics (GRO) methods for monitoring underground storage tanks, trihalomethanes, light hydrocarbons
Volatile organic pollutants, Gasoline range organics,Trihalomethanes, Light hydrocarbons
Specifications
Rtx™-502.2 | |
Fused Silica |