missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Restek™ Soap Film Bubble Flowmeter
For flows between 0.1 and 10 cc/min. requires 1mL flowmeter
Brand: Restek™ 20136
Includes: A reservoir bulb, 24 in. (60cm) of 1/4 in. ID tubing, adapter tubes for 1/8 in. tubing and 0.53mm ID capillary columns, and Velcro™ fasteners.
Description
- 50mL flowmeter for flows between 10 and 300cc/min.
Specifications
Bubble Flowmeter | |
10 and 300 cc/min. | |
1.69 oz. | |
50 mL |