missing translation for 'onlineSavingsMsg'
Learn More

Restek™ VICI™ Mat/Sen™ Gas-Specific Purifier Modules

Replaces separate oxygen, moisture, and hydrocarbon traps with one multiple-bed purifier, specific for purifying helium, hydrogen, nitrogen, or air

Brand:  Restek™ 22607

Code : Z3

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 13104782

  • £655.82 / Each
Estimated Shipment: 12-08-2024
to see stock.

Description

Description

Guaranteed to produce gases that are purer than 99.9999%, when supplied with gas of 99.995% purity, and are prepurged with the specified gas to speed conditioning.

  • Reduces gas impurities from ppm to low ppb levels
  • Decreases baseline noise and increase GC/MS sensitivity
  • Prepurged with the specified gas, to shorten downtime
  • Performances is optimized by incorporating a multiple-bed format that progressively lowers concentrations of contaminants at each successive bed

Specifications

Specifications

0.12 in. Fitting
500 mL/min.
Stainless Steel
21 in.
1.5 in.
Air Purifier Module
Reduce gas impurities from ppm to low ppb levels, prepurged with the specified gas to shorten downtime
1000 psi
52.3 cm
3.8 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers