missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Retention Gap Kits

Install a retention gap/guard column to your analytical column with a single kit.

£28.36 - £58.86

Specifications

For Use With Guard Column (GC)
For Use With (Application) GC
View More Specs

Products 8
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
10506195
View Documents
Thermo Scientific™
260G401P
Tubing; 0.53 mm ID x 1 m (Siltek) N/A N/A N/A
10391435
View Documents
Thermo Scientific™
260G495P
Tubing; 0.25 mm ID x 5 m N/A N/A N/A
10006264
View Documents
Thermo Scientific™
260G496P
Tubing; 0.32 mm ID x 5 m N/A N/A N/A
10301165
View Documents
Thermo Scientific™
260G498P
Tubing; 0.32 mm ID x 2 m
£41.51
Each
Estimated Shipment: 28-04-2025
to see stock.
Add to basket
 
10341335
View Documents
Thermo Scientific™
260G499P
Tubing; 0.53 mm ID x 2 m N/A N/A N/A
10429244
View Documents
Thermo Scientific™
260G500P
Tubing; 0.53 mm ID x 5 m N/A N/A N/A
10691206
View Documents
Thermo Scientific™
260RG497
Retention Gap Kit; 0.22 to 0.32 mm I.D., 2 m N/A N/A N/A
10772846
View Documents
Thermo Scientific™
260RG499
Retention Gap Kit; 0.53 mm I.D., 2 m N/A N/A N/A
Description

Description

Use Thermo Scientific™ Retention Gap Kits to conveniently install a retention gap/guard column to your analytical column. Each kit includes deactivated silica tubing, a miniunion, and five ferrules.

Includes:

  • Length of deactivated fused silica tubing
  • Mini-union
  • Five ferrules
Specifications

Specifications

Guard Column (GC)
GC
Videos
SDS
Documents

Documents

Product Certifications

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Retention Gap Kits

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.