missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Retort Stand Base

Get optimum results using this acid resistant textured acrylic remote stand. Fisherbrand™ Retort Base Stands, are one of the most widely used instruments in experiments.

£46.84 - £67.97

Specifications

Thread Size 10 mm
Finish Pressed Steel
Quantity 1 Ea.
View More Specs

Products 4
Product Code Brand Length (Metric) Price Quantity & Availability  
Product Code Brand Length (Metric) Price Quantity & Availability  
11558092
View accessory products View Documents
Fisherbrand™
6891/SB
200 mm
£46.84
Each
In Stock
to see stock.
Edge
Add to basket
 
11568092
View accessory products View Documents
Fisherbrand™
6892/SB
250 mm
£54.40
Each
In Stock
to see stock.
Edge
Add to basket
 
11578092
View accessory products View Documents
Fisherbrand™
6893/SB
315 mm
£67.97
Each
In Stock
to see stock.
Edge
Add to basket
 
11548092
View accessory products View Documents
Fisherbrand™
6890/SB
160 mm
£48.57
Each
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

  • Pressed steel finished in acid resistant textured acrylic paint
  • Threaded M10 x 1.5mm
  • 10mm threaded holes
  • Single hole stand has a hole centrally located on the width
  • Double hole stand has one hole centrally located on the width and one centrally located along the length
  • Commonly used in filtration, heating experiment
  • One of the most widely used instruments in experiments
  • Simple apparatus for laboratory chemical reactions
Specifications

Specifications

10 mm
Pressed Steel
1 Ea.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Retort Stand Base

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.