Learn More
Brady™ Retro Reflective All Weather Label Tape for M6/M7 Printers
Highly reflective material shines brightly when struck by light which is ideal for applications where high visibility is critical.
Brand:  Brady™ 174130
 View more versions of this product
Product Code. 17828744
  • £254.00 / Each
Estimated Shipment: 04-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ideal for labeling job site tools, hard hats, utility enclosures, poles and pedestals, rooftop equipment, racks and shelves
  • Retroreflective sheeting and pressure sensitive acrylic adhesive meet Federal Spec L-S-300 for Reflectivity
  • Average outdoor durability is 4 to 6 years
  • Service temperature of -40°C to 70°C (-40°F to 158°F)
  • Made of B-584 plastic material
Specifications
Specifications
Not Autoclavable
Permanent Acrylic
Yellow
Continuous
Single-side Printable
Labels
Component and Equipment Labeling, Facility Identification, General Identification, Safety Identification
No
Rectangle
-40°C to +70°C
0.14 mm
30 ft.
2 in.
ASTM D 2979, Fed. Spec. L-S-300 (Class 1)
None
Gloss
Plastic
Thermal Transfer
Non-sterile
M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter)
30 ft.
-40°F to +158°F
0.0055 in.
30 ft. x 2 in.
9.14 m
50.8 mm