Learn More
RETSCH ZM200 Ultra Centrifugal Mill: Distance Sieves
For temperature-sensitive materials
Brand:  RETSCH 03.647.0253
 View more versions of this product
Product Code. 11904252
  • £506.00 / Each
Estimated Shipment: 17-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Distance Sieve
Stainless Steel
Trapezoid holes
ZM200 ultra centrifugal mil
0.08 mm